Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snap-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373059.3 | NP_001360536.1 | 1 - 2 | 64 | 89 | |
NM_072698.6 | NP_505099.2 | 1 - 2 | 60 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |