Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otpl-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072793.4 | NP_505194.2 | 3 - 4 | 456 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |