Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072843.5 | NP_505244.1 | 6 - 7 | 1769 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |