Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072920.1 | NP_505321.1 | 4 - 5 | 622 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |