Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glo-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072991.5 | NP_505392.3 | 7 - 8 | 1283 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |