Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330875.4 | NP_001317742.1 | 10 - 11 | 4005 | 132 | |
NM_001330876.2 | NP_001317741.1 | 10 - 11 | 3999 | 132 | ||
NM_001330877.3 | NP_001317812.1 | 4 - 5 | 1016 | 132 | ||
NM_001330878.3 | NP_001317813.1 | 4 - 5 | 1010 | 132 | ||
NM_001330879.3 | NP_001317814.1 | 3 - 4 | 899 | 132 | ||
NM_001330880.3 | NP_001317815.1 | 3 - 4 | 893 | 132 | ||
NM_001330881.3 | NP_001317816.1 | 2 - 3 | 851 | 132 | ||
NM_001373073.2 | NP_001359965.1 | 11 - 12 | 3926 | 132 | ||
NM_001380727.2 | NP_001367720.1 | 10 - 11 | 3999 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |