Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028557.3 | NP_001023728.1 | 8 - 9 | 1441 | 118 | |
NM_001028558.3 | NP_001023729.1 | 7 - 8 | 1330 | 118 | ||
GenBank mRNA | AY532645.1 | - | 8 - 9 | 1432 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |