Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073215.7 | NP_505616.2 | 8 - 9 | 1087 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |