Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073228.6 | NP_505629.2 | 6 - 7 | 1299 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |