Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 20 - 21 | 3418 | 98 | |
NM_001269276.3 | NP_001256205.1 | 11 - 12 | 1803 | 98 | ||
NM_001269277.3 | NP_001256206.1 | 9 - 10 | 1346 | 98 | ||
NM_001307803.4 | NP_001294732.1 | 9 - 10 | 1386 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |