Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083273.4 | NP_001076742.1 | 2 - 3 | 1013 | 64 | |
NM_001083275.4 | NP_001076744.1 | 1 - 2 | 108 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |