Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083273.4 | NP_001076742.1 | 3 - 4 | 1101 | 84 | |
NM_001083275.4 | NP_001076744.1 | 2 - 3 | 196 | 84 | ||
NM_001356738.3 | NP_001343841.1 | 4 - 5 | 484 | 84 | ||
NM_001356739.4 | NP_001343842.1 | 2 | 206 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |