Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073346.3 | NP_505747.2 | 4 - 5 | 501 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |