Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129546.4 | NP_001123018.1 | 4 - 5 | 568 | 200 | |
NM_073439.3 | NP_505840.1 | 6 - 7 | 842 | 200 | ||
GenBank mRNA | AY008124.1 | - | 6 - 7 | 724 | 200 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |