Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073500.7 | NP_505901.2 | 5 - 6 | 817 | 76 | |
GenBank mRNA | AB044562.1 | - | 5 - 6 | 798 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |