Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073505.8 | NP_505906.2 | 6 - 7 | 1075 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |