Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdz-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073520.3 | NP_505921.1 | 6 - 7 | 1058 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |