Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H12C20.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383409.1 | 7 - 8 | 3359 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |