Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269419.3 | NP_001256348.1 | 5 - 6 | 720 | 70 | |
NM_001269420.3 | NP_001256349.1 | 2 - 3 | 404 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |