Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073560.6 | NP_505961.1 | 1 - 2 | 144 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |