Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073606.5 | NP_506007.1 | 4 - 5 | 520 | 103 | |
NR_001507.1 | 4 - 5 | 448 | 103 | |||
GenBank mRNA | AY557405.1 | - | 4 - 5 | 477 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |