Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269462.4 | NP_001256391.1 | 9 - 10 | 2644 | 62 | |
NM_001269463.3 | NP_001256392.1 | 9 - 10 | 2629 | 62 | ||
NM_001269464.3 | NP_001256393.1 | 2 - 3 | 511 | 62 | ||
GenBank mRNA | AY275181.1 | - | 9 - 10 | 2623 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |