Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073646.7 | NP_506047.5 | 10 - 11 | 1726 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |