Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073656.2 | NP_506057.1 | 1 - 2 | 186 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |