Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129505.5 | NP_001122977.1 | 5 - 6 | 1557 | 114 | |
NM_073662.6 | NP_506063.2 | 13 - 14 | 2913 | 114 | ||
NM_073663.6 | NP_506064.1 | 4 - 5 | 1685 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |