Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129505.5 | NP_001122977.1 | 1 - 2 | 127 | 64 | |
NM_073662.6 | NP_506063.2 | 9 - 10 | 1483 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |