Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073761.7 | NP_506162.1 | 9 - 10 | 2661 | 94 | |
GenBank mRNA | U20168.1 | - | 9 - 10 | 2683 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |