Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cky-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129377.5 | NP_001122849.1 | 6 - 7 | 1511 | 111 | |
GenBank mRNA | AB096001.1 | - | 6 - 7 | 1508 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |