Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269521.2 | NP_001256450.1 | 4 - 5 | 455 | 128 | |
NM_001269522.3 | NP_001256451.1 | 5 - 6 | 663 | 128 | ||
GenBank mRNA | AY008135.1 | - | 4 - 5 | 455 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |