Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bus-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047697.5 | NP_001041162.1 | 3 - 4 | 515 | 149 | |
NR_003174.1 | 3 - 4 | 561 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |