Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nid-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269536.4 | NP_001256465.1 | 6 - 7 | 1330 | 64 | |
NM_001269537.3 | NP_001256466.1 | 6 - 7 | 1336 | 64 | ||
NM_001269538.3 | NP_001256467.1 | 6 - 7 | 1336 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |