Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: him-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073876.4 | NP_506277.1 | 8 - 9 | 2555 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |