Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073889.2 | NP_506290.1 | 2 - 3 | 461 | 103 | |
GenBank mRNA | AY008142.1 | - | 2 - 3 | 459 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |