Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269578.3 | NP_001256507.1 | 6 - 7 | 1177 | 101 | |
NM_001269579.3 | NP_001256508.1 | 2 - 3 | 406 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |