Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nrf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074023.8 | NP_506424.2 | 7 - 8 | 1344 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |