Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stdh-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074047.4 | NP_506448.2 | 2 - 3 | 500 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |