Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380809.2 | NP_001366835.1 | 2 - 3 | 224 | 129 | |
GenBank mRNA | AF059608.1 | - | 1 - 2 | 84 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |