Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380809.2 | NP_001366835.1 | 4 - 5 | 597 | 94 | |
GenBank mRNA | AF059608.1 | - | 3 - 4 | 457 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |