Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ent-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074120.4 | NP_506521.2 | 2 - 3 | 222 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |