Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ent-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074120.4 | NP_506521.2 | 6 - 7 | 861 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |