Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129446.4 | NP_001122918.1 | 10 - 11 | 2040 | 66 | |
NR_070019.1 | 11 - 12 | 1835 | 66 | |||
NR_070020.1 | 10 - 11 | 1712 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |