Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269741.1 | NP_001256670.1 | 2 - 3 | 131 | 93 | |
NM_001269742.2 | NP_001256671.1 | 8 - 9 | 1130 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |