Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074518.3 | NP_506919.2 | 1 - 2 | 199 | 89 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |