Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074553.1 | NP_506954.1 | 2 - 3 | 636 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |