Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074567.4 | NP_506968.1 | 1 - 2 | 204 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |