Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074734.2 | NP_507135.2 | 1 - 2 | 144 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |