Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074744.7 | NP_507145.2 | 2 - 3 | 1972 | 79 | |
GenBank mRNA | AF515833.1 | - | 2 - 3 | 483 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |