Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074854.5 | NP_507255.2 | 5 - 6 | 585 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |