Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029048.6 | NP_001024219.1 | 7 - 8 | 1110 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |