Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ani-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075413.4 | NP_507814.1 | 5 - 6 | 1505 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |